Läs mer
Clomiphene Hydrochloride (E/Z Mixture), TRC
Produktkod. 30136932
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136932 5 g
30134443 25 g
30012480 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136932 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5870555G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57049-00-0
Molekylformel C26 H28 Cl N O . Cl H
Molekylvikt (g/mol) 442.42
Synonym Ethanamine, 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, hydrochloride (1:1), Ethanamine, 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, hydrochloride (9CI), Clomiphene hydrochloride, Clomifene Hydrochloride
IUPAC-namn 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine;hydrochloride
LEDER Cl.CCN(CC)CCOc1ccc(cc1)C(=C(Cl)c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Clomiphene hydrochloride (EZ mixture)
Kvantitet 5 g
InChI formel InChI=1S/C26H28ClNO.ClH/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;/h5-18H,3-4,19-20H2,1-2H3;1H
Formel vikt 441.1626