missing translation for 'onlineSavingsMsg'
Läs mer

Clomipramine N-Glucuronide(>85%), TRC

Produktkod. 30353119
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353119 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30353119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58707010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Clomipramine N-Glucuronide
CAS 165602-83-5
Molekylformel C25 H31 Cl N2 O6
Kvantitet 10 mg
Synonym 3-Chloro-N-β-D-glucopyranuronosyl-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanaminium Inner Salt
InChI formel InChI=1S/C25H32ClN2O6/c1-28(2,24-22(31)20(29)21(30)23(34-24)25(32)33)13-5-12-27-18-7-4-3-6-15(18)8-9-16-10-11-17(26)14-19(16)27/h3-4,6-7,10-11,14,20-24,29-32H,5,8-9,12-13H2,1-2H3/q-1/p-1/t20-,21-,22+,23-,24+/m1/s1
LEDER C[N](C)(CCCN1c2ccccc2CCc3ccc(Cl)cc13)[C@H]4O[C@H]([C@H](O)[C@@H](O)[C@@H]4O)C(=[O-])[O-]
Molekylvikt (g/mol) 490.976
Formel vikt 490.188
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.