Läs mer
Clomipramine N-Oxide, TRC
Produktkod. 30355117
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355117 10 mg
30364720 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30355117 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58706010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clomipramine N-Oxide
CAS 14171-67-6
Molekylformel C19 H23 Cl N2 O
Kvantitet 10 mg
Synonym Clomipramine N-Oxide
InChI formel InChI=1S/C19H23ClN2O/c1-22(2,23)13-5-12-21-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)21/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3
LEDER C[N+](C)([O-])CCCN1c2ccccc2CCc3ccc(Cl)cc13
IUPAC-namn 3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine oxide
Molekylvikt (g/mol) 330.85
Formel vikt 330.1499