Läs mer
Clonidine EP Impurity B (Clonidine USP Related Compound A), Mikromol™
Produktkod. 30051864
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30051864 25 mg
1 options
Produktkod. 30051864 Leverantör Mikromol Leverantörsnummer MM0983.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Clonidine EP Impurity B (Clonidine USP Related Compound A)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Acetyl-2-[(2,6-dichlorophenyl)amino]-4,5-dihydro-1H-imidazole
CAS 54707-71-0
Analyt- eller komponentnamn 1-Acetyl-2-[(2,6-dichlorophenyl)amino]-4,5-dihydro-1H-imidazole
Frakt skick Room Temperature
Molekylformel C11 H11 Cl2 N3 O
Certifikat och efterlevnad ISO 17025
Synonym Clonidine Hydrochloride Imp. B (EP), Clonidine Imp. B (EP), Clonidine USP Related Compound A, Clonidine USP RC A, 1-Acetyl-2-[(2,6-dichlorophenyl)amino]-4,5-dihydro-1H-imidazole, Clonidine Related Compound A, Clonidine Hydrochloride Impurity B, Clonidine Impurity B
InChI formel InChI=1S/C11H11Cl2N3O/c1-7(17)16-6-5-14-11(16)15-10-8(12)3-2-4-9(10)13/h2-4H,5-6H2,1H3,(H,14,15)
LEDER CC(=O)N1CCN=C1Nc2c(Cl)cccc2Cl
IUPAC-namn 1-[2-(2,6-dichloroanilino)-4,5-dihydroimidazol-1-yl]ethanone
Molekylvikt (g/mol) 272.13
Kvantitet 25 mg
Formel vikt 271.0279 g/mol