Läs mer
Clonquitocet-methyl, TRC
Produktkod. 30354265
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30354265 500 mg
1 options
Produktkod. 30354265 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC436790500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clonquitocet-methyl
CAS 4367-49-1
Molekylformel C12 H10 Cl N O3
Kvantitet 500 mg
Synonym (5-Chloro-quinolin-8-yloxy)-acetic Acid Methyl Ester, Cloquintocet-methyl, Methyl 5-Chloro-8-hydroxyquinolyloxyacetate
InChI formel InChI=1S/C12H10ClNO3/c1-16-11(15)7-17-10-5-4-9(13)8-3-2-6-14-12(8)10/h2-6H,7H2,1H3
LEDER COC(=O)COc1ccc(Cl)c2cccnc12
IUPAC-namn methyl 2-(5-chloroquinolin-8-yl)oxyacetate
Molekylvikt (g/mol) 251.666
Formel vikt 251.035