Läs mer
Clopidogrel Alcohol, Mikromol™
Produktkod. 30461463
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461463 4 x 25 mg
17920496 100 mg
17901686 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461463 Leverantör Mikromol Leverantörsnummer MM0455.0900254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Clopidogrel Alcohol, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Clopidogrel Alcohol
CAS 1227294-01-0
Analyt- eller komponentnamn Clopidogrel Alcohol
Frakt skick Room Temperature
Molekylformel C15 H16 Cl N O S
Certifikat och efterlevnad ISO 17025
Synonym Clopidogrel Alcohol
InChI formel InChI=1S/C15H16ClNOS/c16-13-4-2-1-3-12(13)14(10-18)17-7-5-15-11(9-17)6-8-19-15/h1-4,6,8,14,18H,5,7,9-10H2/t14-/m1/s1
Lösningstyp Neat
LEDER OC[C@@H](N1CCc2sccc2C1)c3ccccc3Cl
IUPAC-namn (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)ethanol
Molekylvikt (g/mol) 293.81
Kvantitet 4 x 25 mg
Formel vikt 293.0641 g/mol