Läs mer
Clopidogrel Carboxylic Acid Hydrochloride, TRC
Produktkod. 17940565
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17940565 50 mg
17950565 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17940565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58723550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144750-42-5
Molekylformel C15 H14 Cl N O2 S . Cl H
Molekylvikt (g/mol) 344.26
Synonym Clopidogrel (free acid) hydrochloride, Thieno[3,2-c]pyridine-5(4H)-acetic acid, α-(2-chlorophenyl)-6,7-dihydro-, hydrochloride (1:1), (αS)-, Thieno[3,2-c]pyridine-5(4H)-acetic acid, α-(2-chlorophenyl)-6,7-dihydro-, hydrochloride, (αS)- (9CI), (+)-(S)-(o-chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid, hydrochloride
IUPAC-namn (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid;hydrochloride
LEDER Cl.OC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl
Specifikationer
Kemiskt namn eller material Clopidogrel carboxylic acid hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C15H14ClNO2S.ClH/c16-12-4-2-1-3-11(12)14(15(18)19)17-7-5-13-10(9-17)6-8-20-13;/h1-4,6,8,14H,5,7,9H2,(H,18,19);1H/t14-;/m0./s1
Formel vikt 343.0201
Procent renhet >95%