Läs mer
Clopidogrel Carboxylic Acid [Methyl (R)-o-chloromandelate] Ester, TRC
Produktkod. 30357374
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357374 1 mg
30366466 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30357374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5873201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1421283-60-4
Molekylformel C24H21Cl2NO4S
Molekylvikt (g/mol) 490.4
Synonym Clopidogrel Impurity D, Clopidogrel Ethyl Ester Impurity
IUPAC-namn [(1R)-1-(2-chlorophenyl)-2-methoxy-2-oxoethyl] (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate
LEDER COC(=O)[C@H](OC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl)c4ccccc4Cl
Specifikationer
Kemiskt namn eller material Clopidogrel Carboxylic Acid [Methyl (R)-o-chloromandelate] Ester
Kvantitet 1 mg
InChI formel InChI=1S/C24H21Cl2NO4S/c1-30-24(29)22(17-7-3-5-9-19(17)26)31-23(28)21(16-6-2-4-8-18(16)25)27-12-10-20-15(14-27)11-13-32-20/h2-9,11,13,21-22H,10,12,14H2,1H3/t21-,22+/m0/s1
Formel vikt 489.0568