Läs mer
Clorprenaline Hydrochloride, TRC
Produktkod. 30363416
Change view
Kvantitet:
500 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30363416 500 mg -
30350330 2.5 g 2.5g
30350614 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363416 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC427955500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6933-90-0
Molekylformel C11 H16 Cl N O . Cl H
Molekylvikt (g/mol) 250.16
Synonym Benzenemethanol, 2-chloro-α-[[(1-methylethyl)amino]methyl]-, hydrochloride (9CI), Benzyl alcohol, o-chloro-α-[(isopropylamino)methyl]-, hydrochloride (6CI,7CI,8CI), Chlorprenaline hydrochloride, Clorprenaline hydrochloride, Isoprophenamine hydrochloride, N-[2-(2-Chlorophenyl)-2-hydroxyethyl]propan-2-aminium chloride, NSC 334693
IUPAC-namn 1-(2-chlorophenyl)-2-(propan-2-ylamino)ethanol;hydrochloride
LEDER Cl.CC(C)NCC(O)c1ccccc1Cl
Specifikationer
Kemiskt namn eller material Clorprenaline Hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C11H16ClNO.ClH/c1-8(2)13-7-11(14)9-5-3-4-6-10(9)12;/h3-6,8,11,13-14H,7H2,1-2H3;1H
Formel vikt 249.0687