Learn More
Closantel 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30314230
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30314230 1 mL 1mL
1 options
Product Code. 30314230 Supplier Dr. Ehrenstorfer Supplier No. DREA11691500AL100
 Restricted Item
Beräknad leverans: 30-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Closantel
CAS 57808-65-8
Analyte or Component Names Closantel
Shipping Condition Room Temperature
Molecular Formula C22 H14 Cl2 I2 N2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym N-[5-chloro-4-[(4-chlorophenyl)cyanomethyl]-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide, Closantel, Clozantin, NSC 335306, Seponver, Zycloz
InChI Formula InChI=1S/C22H14Cl2I2N2O2/c1-11-6-15(17(10-27)12-2-4-13(23)5-3-12)18(24)9-20(11)28-22(30)16-7-14(25)8-19(26)21(16)29/h2-9,17,29H,1H3,(H,28,30)
SMILES Cc1cc(C(C#N)c2ccc(Cl)cc2)c(Cl)cc1NC(=O)c3cc(I)cc(I)c3O
IUPAC Name N-[5-chloro-4-[(4-chlorophenyl)-cyanomethyl]-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 663.07
Quantity 1 mL
Formula Weight 661.8522 g/mol