Läs mer
Cloticasone Propionate, TRC
Produktkod. 30134064
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134064 100 mg
30133440 50 mg
17974035 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134064 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC369160100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80486-69-7
Molekylformel C25 H31 Cl F2 O5 S
Molekylvikt (g/mol) 517.03
Synonym CCI 18773
IUPAC-namn [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-17-(chloromethylsulfanylcarbonyl)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
LEDER CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCCl
Specifikationer
Kemiskt namn eller material Cloticasone Propionate
Kvantitet 100 mg
InChI formel InChI=1S/C25H31ClF2O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1
Formel vikt 516.1549