Learn More
CLPB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558623
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GKTIDCKDAI FIMTSNVASD EIAQHALQLR QEALEMSRNR IAENLGDVQI SDKITISKNF KENVIRPILK AHFRRDEFLG RINEIVYFLP FCHSELIQLV NKELNFWA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
CLPB Polyclonal antibody specifically detects CLPB in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
CLPB | |
Polyclonal | |
Unconjugated | |
Clpb | |
AL118244; ANKCLB; ankyrin-repeat containing bacterial clp fusion; caseinolytic mitochondrial matrix peptidase chaperone subunit B; caseinolytic peptidase B; caseinolytic peptidase B protein homolog; CLPB; ClpB caseinolytic peptidase B; ClpB caseinolytic peptidase B homolog; ClpB caseinolytic peptidase B homolog (E. coli); ClpB homolog, mitochondrial AAA ATPase chaperonin; HSP78; MEGCANN; MGCA7; Skd3; suppressor of K+ transport defect 3; suppressor of potassium transport defect 3; testicular secretory protein Li 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81570 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H078 | |
Clpb | |
Recombinant protein corresponding to Human CLPB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only