Learn More
CLS-VF, MP Biomedicals™
Cell Lysis/DNA solubilizing solution for plant tissues extractions.
Brand: MP Biomedicals 116540402
Additional Details : Weight : 0.01000kg
Description
CLS-VF is a cell Lysis/DNA solubilizing 90 mL solution for vegetation. For plant tissues, CLS-VF is used in conjunction with a Protein Precipitation Solution (PPS).Specifications
90 mL | |
Liquid |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.