Learn More
CLS-VF, MP Biomedicals™
Cell Lysis/DNA solubilizing solution for plant tissues extractions.
Brand:  MP Biomedicals 116540402
Additional Details : Weight : 0.01000kg
Product Code. 11452390
  • 1261.40 SEK / 90mL
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
CLS-VF is a cell Lysis/DNA solubilizing 90 mL solution for vegetation. For plant tissues, CLS-VF is used in conjunction with a Protein Precipitation Solution (PPS).
Specifications
Specifications
90 mL
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.