missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTB Mouse anti-Human, Porcine, Clone: 1F9C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66270-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTB | |
Monoclonal | |
1.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09497 | |
CLTB | |
CLTB Fusion Protein Ag24347 | |
20 μL | |
Primary | |
Porcine, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
1F9C2 | |
Unconjugated | |
CLTB | |
Clathrin light chain B, clathrin, light chain (Lcb), CLTB, LCB | |
Mouse | |
Protein G | |
RUO | |
100525860, 1212 | |
-20°C | |
Liquid |