missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTB Mouse anti-Human, Porcine, Clone: 1F9C2, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CLTB |
---|---|
Clone | 1F9C2 |
Concentration | 1.32 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827995
|
Proteintech
66270-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817995
|
Proteintech
66270-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTB | |
1.32 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Clathrin light chain B, clathrin, light chain (Lcb), CLTB, LCB | |
CLTB | |
IgG1 | |
Protein G | |
Antibody |
1F9C2 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Porcine, Human | |
P09497 | |
100525860, 1212 | |
CLTB Fusion Protein Ag24347 | |
Primary | |
-20°C | |
CLTB |