missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMPK1 Mouse anti-Human, Unconjugated, Clone: 2D11G7, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CMPK1 |
---|---|
Clone | 2D11G7 |
Concentration | 500 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840196
|
Proteintech
67519-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830196
|
Proteintech
67519-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CMPK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CMPK1. This antibody recognizes human antigen. The CMPK1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
CMPK1 | |
500 μg/mL | |
Monoclonal | |
Mouse | |
51727 | |
Primary |
2D11G7 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG2b |