missing translation for 'onlineSavingsMsg'
Learn More

CMTM8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15039-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824014

  • 1236.20 SEK / 20µL
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown.
Specifications

Specifications

CMTM8
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IZV2, Q9CZR4
CKLFSF8, CKLFSF8 V2, CMTM8
Rabbit
IgG
20 μL
-20°C
Primary
152189, 301045, 70031
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CMTM8
Liquid
Cmtm8
CMTM8 Fusion Protein Ag6776
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers