Beskrivning
CNBD2 Polyclonal antibody specifically detects CNBD2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifikationer
Specifikationer
| Antigen | CNBD2 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | DyLight 488 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | chromosome 20 open reading frame 152, dJ954P9.1, hypothetical protein LOC140894, MGC138880 |
| Värd art | Rabbit |
| Immunogen | chromosome 20 open reading frame 152. (Uniprot# Q96M20) |
| Reningsmetod | Immunogen affinity purified |
| Kvantitet | 0.1 mL |
| Visa mer |