missing translation for 'onlineSavingsMsg'
Learn More

CNDP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17759-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827724

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
Specifications

Specifications

CNDP1
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q66HG3, Q8BUG2, Q96KN2
Beta Ala His dipeptidase, Carnosine dipeptidase 1, CN1, CNDP dipeptidase 1, CNDP1, CPGL2, HsT2308, Serum carnosinase
Rabbit
IgG
150 μL
-20°C
Primary
307212, 338403, 84735
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CNDP1
Liquid
Cndp1
CNDP1 Fusion Protein Ag12127
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers