missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CNOT2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886613
|
Proteintech
10313-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16876613
|
Proteintech
10313-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CCR4-NOT complex functions as general transcription regulation complex. Subunit of the CCR4-NOT core complex that contains CHAF1A, CHAF1B, CNOT1, CNOT2, CNOT3, CNOT4, CNOT6 and CNOT8. 5 isoforms of the human protein are produced by alternative splicing.Specifications
CNOT2 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8C5L3, Q9NZN8 | |
299805, 4848, 72068 | |
CNOT2 Fusion Protein Ag0347 | |
Primary | |
-20°C | |
CNOT2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCR4 associated factor 2, CDC36, CNOT2, HSPC131, MSTP046, NOT2, NOT2H | |
CNOT2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |