missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10313-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The CCR4-NOT complex functions as general transcription regulation complex. Subunit of the CCR4-NOT core complex that contains CHAF1A, CHAF1B, CNOT1, CNOT2, CNOT3, CNOT4, CNOT6 and CNOT8. 5 isoforms of the human protein are produced by alternative splicing.Specifications
CNOT2 | |
Polyclonal | |
Unconjugated | |
CNOT2 | |
CCR4 associated factor 2, CDC36, CNOT2, HSPC131, MSTP046, NOT2, NOT2H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
299805, 4848, 72068 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C5L3, Q9NZN8 | |
CNOT2 | |
CNOT2 Fusion Protein Ag0347 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |