missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CNTFR Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518622
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is predicted to react with canine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a hematopoeitin/interferon-class receptor belonging to the cytokine superfamily of receptors. The encoded gene product represents the CNTF-specific alpha subunit of a heterotrimer forming the CNTF receptor complex, which also includes LIFR and gp130. The receptor is attached to the membrane by a glycosyl-phosphatidylinositol linkage and contains an immunoglobulin-like C2-type domain and a fibronectin type-III domain. Signal transduction requires that CNTF bind first to this alpha component, which permits the recruitment of gp130 and LIFR beta to form the tripartite receptor complex. Signal transduction stimulates gene expression, cell survival or differentiation in a variety of neuronal cell types. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified.
Specifikationer
| CNTFR | |
| Polyclonal | |
| Unconjugated | |
| Cntfr | |
| ciliary neurotrophic factor receptor; ciliary neurotrophic factor receptor alpha; ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; Cntfr; CNTFR alpha; CNTFRALPHA; CNTFR-alpha; MGC1774 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 12804 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O88507 | |
| Cntfr | |
| Synthetic peptide sequence (CRSNTYPKGFY) corresponding to the internal amino acids of CNTFR (aa 116-127). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Product Title
Spot an opportunity for improvement?Share a Content Correction