Learn More
Cobalt(II) acetate, anhydrous, 98+%, Thermo Scientific Chemicals
Catalog Number p-7026908
Click to view available options
Quantity:
50 g
250 g
71-48-7
C4H6CoO4
177.02
MFCD00008689
QAHREYKOYSIQPH-UHFFFAOYSA-L
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
6277
CHEBI:85138
cobalt(2+);diacetate
[Co++].CC([O-])=O.CC([O-])=O
71-48-7
C4H6CoO4
177.02
MFCD00008689
QAHREYKOYSIQPH-UHFFFAOYSA-L
cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
6277
CHEBI:85138
cobalt(2+);diacetate
[Co++].CC([O-])=O.CC([O-])=O
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.
Solubility
Soluble in water, alcohol and dilute acids.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 298°C (decomposition)
Odor Odorless
Linear Formula Co(OOCCH3)2
Quantity 50 g
UN Number UN3077
Sensitivity Hygroscopic
Merck Index 14,2433
Solubility Information Soluble in water,alcohol and dilute acids.
Formula Weight 177.02
Percent Purity ≥98%
Chemical Name or Material Cobalt(II) acetate, anhydrous