Läs mer
Cobalt(II) Chloride Hydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Cobalt(II) Chloride Hydrate |
| CAS | 69098-14-2 |
| Molekylformel | Cl2Co •xH2O |
| Kvantitet | 25 g |
| Synonym | Cobalt Chloride Hydrate, Cobalt Dichloride Hydrate, Cobalt Dichloride Hydrate (CoCl2), Cobalt(2+) Chloride Hydrate, Cobalt(II) Chloride Hydrate, Cobaltous Chloride Hydrate, Cobaltous Dichloride Hydrate, Dichlorocobalt Hydrate, NSC 51149 |
| InChI formel | InChI=1S/2ClH.Co.H2O/h2*1H;;1H2/q;;+2;/p-2 |
| LEDER | Cl[Co]Cl |
| IUPAC-namn | dichlorocobalt;hydrate |
| Molekylvikt (g/mol) | 129.841801 |