Learn More
Cobalt(II) Nitrate Hexahydrate, Supelco™
Product Code. 15857120
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15857120 100 g 100g
15854270 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15857120 Supplier Supelco Supplier No. 1025360100
 Restricted Item
Beräknad leverans: 26-11-2026
For analysis EMSURE™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10026-22-9
Specifications
Melting Point 57°C
Quantity 100 g
Molecular Formula Co(NO3)2 · 6 H2O
MDL Number MFCD00149647
Synonym Cobalt(II) nitrate hexahydrate, Cobalt(II) nitrate hexahydrate
Molecular Weight (g/mol) 291.03
Grade Research
Density 1.87 g/cm3 at 20°C
Chemical Name or Material Cobalt(II) Nitrate Hexahydrate