Learn More
Alfa Aesar™ Cobalt(II) oxide, 99.998% (metals basis)
Brand:  Alfa Aesar™ 045907.06
Additional Details : CAS Number : 1307-96-6 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity 5g
 View more versions of this product
Product Code. 15492547
  • 1426.94 SEK / 5g
Estimated Shipment date
from Supplier 28-02-2018
Add to basket
Specifications
Chemical Identifiers
Cobalt (II) oxide
74.93
UN2811
1307-96-6
MFCD00016031
14,2446
Specifications
Cobalt (II) oxide
Crystals
(metals basis)
MFCD00016031
14,2446
6.45g/mL
Air Sensitive
1935°C
1307-96-6
99.998%
74.93
UN2811
CoO, may contain up to 10% Co3O4
3 to 7mm
Green-brown
5g