Läs mer
Cocarboxylase Tetrahydrate(>90%), TRC
Produktkod. 30137002
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137002 50 mg
30134139 100 mg
30008357 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137002 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC63351550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68684-55-9
Molekylformel C12H18N4O7P2S . 4H2O
Molekylvikt (g/mol) 496.37
Synonym Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)-, inner salt, hydrate (1:4), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-3,5-dioxa-4,6-diphosphahex-1-yl)-, inner salt, P,P'-dioxide, tetrahydrate, Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)-, inner salt, tetrahydrate (9CI), Cocarboxylase tetrahydrate, Thiamine pyrophosphate (inner salt) tetrahydrate
IUPAC-namn [2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethoxy-hydroxyphosphoryl] hydrogen phosphate;tetrahydrate
LEDER O=P(O)(O)OP(=O)([O-])OCCC=1SC=[N+](C1C)CC2=CN=C(N=C2N)C.O
Specifikationer
Kemiskt namn eller material Cocarboxylase Tetrahydrate
Kvantitet 50 mg
InChI formel InChI=1S/C12H18N4O7P2S.4H2O/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;;;;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);4*1H2
Formel vikt 496.0794