Läs mer
DiscoverX Coelenterazine h
Beskrivning
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 10mg of dry powder.
Specifikationer
Specifikationer
| Kvantitet | 10 mg |
| Produkttyp | Biochemical Reagent |
| Målinträde | Calcium |