Läs mer
Coenzyme A Trihydrate, Free Acid, Thermo Scientific™
Produktkod. 15895268 Handla allt Thermo Scientific Produkter
Change view
Kvantitet:
50 mg
100 mg
250 mg
1 g
Förpackning:
Plastic bottle
Förpackningsstorlek:
100mg
1g
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackning unitSize
15895268 50 mg Plastic bottle 50mg
15875268 100 mg Plastic bottle 100mg
15885268 250 mg Plastic bottle 250mg
15805118 1 g Plastic bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 15895268 Leverantör Thermo Scientific Leverantörsnummer J13787.MF
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85-61-0
Molekylformel C21H36N7O16P3S
Molekylvikt (g/mol) 767.533
MDL-nummer MFCD0679839
InChI-nyckel RGJOEKWQDUBAIZ-RXPJKXIYSA-N
Synonym coenzymea, coenzyme a
PubChem CID 133639590
IUPAC-namn [[(2S,3R,4S,5S)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(3R)-3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butyl] hydrogen phosphate
LEDER CC(C)(COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)C(C(=O)NCCC(=O)NCCS)O
Specifikationer
Kemiskt namn eller material Coenzyme A trihydrate
Förpackning Plastic bottle
Kvantitet 50 mg
Beilstein 77809
Formel vikt 821.58
Fysisk form Lyophilized Powder