missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11130-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a component of the conserved oligomeric Golgi (COG) complex which is composed of eight different subunits and is required for normal Golgi morphology and localization. Defects in the COG complex result in multiple deficiencies in protein glycosylation. The protein encoded by this gene is involved in ER-Golgi transport.Specifications
COG3 | |
Polyclonal | |
Unconjugated | |
COG3 | |
COG complex subunit 3, COG3, p94, SEC34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
338337, 361073, 83548 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CI04, Q96JB2 | |
Cog3 | |
COG3 Fusion Protein Ag1610 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |