Läs mer
COH000, MedChemExpress
Produktkod. 16464808
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16464808 10 mg
16447557 5 mg
16445606 100 mg
16401077 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16464808 Leverantör Medchem Express Leverantörsnummer HY114304
MedChemExpress COH000 is an allosteric, covalent and irreversible inhibitor of ubiquitin-like 1-activating enzyme (SUMO-activating enzyme) (E1), with an IC50 of 0.2 μM for SUMOylation in vitro.
COH000 is an allosteric, covalent and irreversible inhibitor of ubiquitin-like 1-activating enzyme (SUMO-activating enzyme) (E1), with an IC50 of 0.2 μM for SUMOylation in vitro.
Specifikationer
Kemiskt namn eller material COH000
CAS 1534358-79-6
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C25H25NO5
Kvantitet 10 mg
Löslighetsinformation DMSO : 35.71 mg/mL (85.13 mM; Need ultrasonic)
LEDER CC(C=C1)=CC=C1C[C@@H](NC2=CC=CC=C2)[C@]34C(C(OC)=O)=C(C(OC)=O)[C@@H](O4)C=C3
Molekylvikt (g/mol) 419.47
Formel vikt 419.47
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death