Läs mer
COL4A3 Antibody (COL4A3/9395R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-44590-20ug
2267.22 SEK valid until 2025-12-16
BÄSTA PRIS på kampanj! Använd kampanjkod: "24090" för att få ditt kampanjpris.
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
COL4A3 Monoclonal antibody specifically detects COL4A3 in Human samples. It is validated for Immunohistochemistry
Specifikationer
| COL4A3 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 1285 | |
| Store at 4⪚C. | |
| IgG κ |
| Immunohistochemistry | |
| COL4A3/9395R | |
| Unconjugated | |
| ATS2, ATS3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), tumstatin | |
| Recombinant fragment (around aa1400-1670) of human COL4A3 protein (Uniprot: P02462, P08572, P29400, P53420). Exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |