missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ COL8A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537109
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody detects endogenous protein at a molecular weight of 73 kDa. Purity is >95% by SDS-PAGE.
The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (types I, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
Specifikationer
| COL8A1 | |
| Polyclonal | |
| Unconjugated | |
| COL8A1 | |
| C3orf7; cell proliferation-inducing protein 41; Col8a1; Col8a-1; Collagen alpha-1(VIII) chain; collagen type VIII alpha 1; collagen VIII, alpha-1 polypeptide; collagen, type VIII, alpha 1; endothelial collagen; procollagen type VIII alpha 1; procollagen, type VIII, alpha 1; smag-64; smooth muscle cell-expressed and macrophage conditioned medium-induced protein smag-64; Vastatin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12837, 1295, 304021 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27658, Q00780 | |
| COL8A1 | |
| A synthetic peptide corresponding to residues in human COL8A1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering