Learn More
Colchicine, MedChemExpress
Product Code. 16461226
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461226 200 mg 200mg
16431816 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16461226 Supplier Medchem Express Supplier No. HY16569
 Restricted Item
Beräknad leverans: 14-12-2026
MedChemExpress Colchicine is a tubulin inhibitor and a microtubule disrupting agent. Colchicine inhibits microtubule polymerization with an IC50 of 3 nM. Colchicine is also a competitive antagonist of the α3 glycine receptors (GlyRs).
Colchicine is a tubulin inhibitor and a microtubule disrupting agent. Colchicine inhibits microtubule polymerization with an IC50 of 3 nM. Colchicine is also a competitive antagonist of the α3 glycine receptors (GlyRs).
Specifications
Chemical Name or Material Colchicine
CAS 64-86-8
Color White
Purity Grade Notes Research
Molecular Formula C22H25NO6
Quantity 200 mg
Solubility Information DMSO : ≥ 48 mg/mL (120.17 mM) ∣H2O : ≥ 33.33 mg/mL (83.44 mM)
SMILES CC(N[C@H](C1=C2)CCC3=CC(OC)=C(OC)C(OC)=C3C1=CC=C(OC)C2=O)=O
Molecular Weight (g/mol) 399.44
Formula Weight 399.44
Percent Purity 98.62%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death