Läs mer
Colesevelam Hydrochloride (Technical Grade), TRC
Produktkod. 30133412
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133412 25 mg
30135083 50 mg
30009549 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133412 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC64009525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 182815-44-7
Molekylformel (C12H27ClN2 • C13H27N • C3H7N • C3H5ClO)x • (HCl)x
Molekylvikt (g/mol) 618.24
Synonym CholestaGel, GT 31-104, GT 31-104HB, Welchol;
IUPAC-namn 1-Hexanaminium, N,N,N-trimethyl-6-(2-propen-1-ylamino)-, chloride (1:1), polymer with 2-(chloromethyl)oxirane, 2-propen-1-amine and N-2-propen-1-yl-1-decanamine, hydrochloride (ACI)
LEDER C=CCNCCCCCC[N+](C)(C)C.C=CCNCCCCCCCCCC.C=CCN.ClCC1CO1.[Cl-].[HCl]
Specifikationer
Kemiskt namn eller material Colesevelam Hydrochloride (Technical Grade)
Kvantitet 25 mg
InChI formel InChI=1S/C13H27N.C12H27N2.C3H5ClO.C3H7N.2ClH/c1-3-5-6-7-8-9-10-11-13-14-12-4-2;1-5-10-13-11-8-6-7-9-12-14(2,3)4;4-1-3-2-5-3;1-2-3-4;;/h4,14H,2-3,5-13H2,1H3;5,13H,1,6-12H2,2-4H3;3H,1-2H2;2H,1,3-4H2;2*1H/q;+1;;;;/p-1