Learn More
Collagen Type III (N-terminal) Rabbit anti-Human, Unconjugated, Clone: 2O1, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80009-1-RR-20UL
Additional Details : Weight : 0.00010kg
Description
The Collagen Type III (N-terminal) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Collagen Type III (N-terminal). This antibody recognizes human antigen. The Collagen Type III (N-terminal) antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Collagen Type III (N-terminal) | |
Monoclonal | |
750 μg/mL | |
Rabbit | |
Primary | |
Human |
Immunohistochemistry, Immunofluorescence | |
2O1 | |
Unconjugated | |
20 μL | |
1281 | |
IgG |