Learn More
STEMCELL Technologies Collagenase B, ACF
Animal component-free collagenase for the digestion of native collagen fibrils
2116.03 SEK - 9691.71 SEK
Specifications
For Use With (Application) Digestion of Native Collagen Fibrils
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17189261
View Documents Promotion Details
STEMCELL Technologies
07438
100 mg
2116.03 SEK
100mg
View alternative products
Save Up to 
»
Estimated Shipment: 10-11-2023
to see stock.
17199261
View Documents Promotion Details
STEMCELL Technologies
07439
1 g
9691.71 SEK
1g
View alternative products
Save Up to 
»
Estimated Shipment: 10-11-2023
to see stock.
Description
Description
  • Collagenase B, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
  • Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
  • Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
  • Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
  • Collagenase B, ACF contains higher collagenase and caseinase levels than Collagenase A and is intended for use in applications where it is necessary to prevent the introduction of potential animal-derived pathogens
Specifications
Specifications
Digestion of Native Collagen Fibrils