missing translation for 'onlineSavingsMsg'
Läs mer

Nordmark Biochemicals Collagenase NB 6, GMP Grade

Produktkod. 11467503
Encompass
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
Förpackningsstorlek:
Varje
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11467503 1 g Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11467503 Leverantör Nordmark Biochemicals Leverantörsnummer 1745801

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Suitable for applications that must meet GMP requirements, such as tissue engineering and transplantation into humans. Nordmark Biochemicals Collagenase NB 6, GMP Grade is a superior grade lyophilized enzyme with sterility according to Ph. Eur. and TSE safety certified.

Collagenases from Clostridium histolyticum are proteolytic enzymes that cleave peptide bonds in the triple helical collagen molecule of human or animal tissue in situ. Nordmark Collagenase NB products are particularly suitable for cell isolation from various tissues and passaging of stem cells.

  • Outstanding quality: Manufactured in compliance with GMP guidelines
  • Ready for use: Sterile according to European Pharmacopeia
  • Exceptionally safe: Virus validation studies and lot testing for abnormal toxicity
  • Reproducibility: Reliable lot-to-lot consistency
  • Regulatory advantage: TSE safety certificate and US DMF available

Specifikationer

Färg Light brown
Kvalitet NB 6, GMP Grade
För användning med (applikation) Isolation of various cell types by tissue dissociation
Förpackning Bag
Löslighet Soluble in common buffers, including water at 300g/L; Dissolves at a concentration of up to 150mg/mL
CAS 9001-12-1
Innehåll och lagring Store at 2°-8°C for one year
Protein Adipose tissue, cartilage, skin, placenta, umbilical cord tissue, lung and for cell culture of embryonic stem cells
Källa Clostridium histolyticum
Kvantitet 1 g
Produkttyp Collagenase (proteolytic enzyme)
Inhibitorer Inhibitors include cysteine or chelating agents (e.g. EDTA)
Form Lyophilized Powder
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.