Learn More
Witeg™ Color Comparing Tubes Acc. to Eggertz
For the colorimetric estimation of carbon and manganese in steel.
Brand: Witeg™ 2 149 050
Additional Details : Weight : 0.06400kg
Description
- With ST 12/21 stopper
- Capacity of 50 mL
- Manufactured from borosilicate glass
Specifications
50 mL | |
Stopper | |
ST 12/21 |
Borosilicate Glass | |
Colorimetric estimation |