Learn More
Thermo Scientific Chemicals ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip
607.07 SEK - 3550.13 SEK
Product Code | Brand | Quantity | pH Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | pH Range | Price | Quantity & Availability | ||||
11323607
|
Thermo Scientific Chemicals
035234.LQ |
1 Unit | 5 to 10 |
698.19 SEK
1 unit |
In Stock
Log in to see stock. |
||||
11313607
|
Thermo Scientific Chemicals
035234.LS |
6 Unit | 5 to 10 |
3384.46 SEK
6 units |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
11303607
|
Thermo Scientific Chemicals
035233.LQ |
1 Unit | 7.5 to 14 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11393597
|
Thermo Scientific Chemicals
035233.LS |
6 Unit | 7.5 to 14 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11333607
|
Thermo Scientific Chemicals
035235.LQ |
1 Unit | 0 to 6 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11343607
|
Thermo Scientific Chemicals
035235.LS |
6 Unit | 0 to 6 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11353607
|
Thermo Scientific Chemicals
035236.LQ |
1 Unit | 2 to 9 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11363607
|
Thermo Scientific Chemicals
035236.LS |
6 Unit | 2 to 9 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11373607
|
Thermo Scientific Chemicals
035237.LQ |
1 Unit | 0 to 14 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11383607
|
Thermo Scientific Chemicals
035237.LS |
6 Unit | 0 to 14 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
ColorpHast™ pH Strips, Universal, four squares with different indicators on each strip, 0-14 pH range is used in analytical laboratory to measure the pH readings in seconds in drinking water, saliva and in chemical reactions. It is utilized for quick and accurate measurement and saves our time and trouble.