Learn More
Fisherbrand™ Comb 1.5mm
Manufactured to the highest standard. Fisherbrand™ Comb 1.5mm is used with electrophoresis systems.
676.00 SEK - 769.00 SEK
Specifications
For Use With (Equipment) | Vertical Gel Electrophoresis |
---|---|
Thickness (Metric) Teeth | 1.5 mm |
Product Code | Brand | No. of Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Price | Quantity & Availability | |||||
11744296
|
Fisherbrand™
|
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11724296
|
Fisherbrand™
|
24 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11789675
|
Fisherbrand™
|
10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Used with vertical electrophoresis systems
Specifications
Vertical Gel Electrophoresis | |
1.5 mm |