Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 2mm Thick
For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11817823
Quantity Price
1 284.35 SEK / Each
10 255.92 SEK / Each
Estimated Shipment
24-05-2021
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Blue
Comb
1 Each
100 μL
Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units
18
2 mm