Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
262.00 SEK
Specifications
Type Comb
Quantity 1 Each
Thickness (Metric) 2 mm
View More Specs
Products 9
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
11823313
View Documents Promotion Details
Fisherbrand™
10 54 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
11853313
View Documents Promotion Details
Fisherbrand™
1 Prep, 1 Marker 720 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
11893313
View Documents Promotion Details
Fisherbrand™
12 45 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
11833323
View Documents Promotion Details
Fisherbrand™
16 32 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
11883323
View Documents Promotion Details
Fisherbrand™
20 27 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
11813333
View Documents Promotion Details
Fisherbrand™
2 Prep, 2 Marker 315 μL x 8
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
11853333
View Documents Promotion Details
Fisherbrand™
25 20 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
11893333
View Documents Promotion Details
Fisherbrand™
4 Prep, 2 Marker 153 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
11813353
View Documents Promotion Details
Fisherbrand™
8 81 μL
262.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Comb
1 Each
2 mm