missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit, 1.5mm thick
For use with HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- Color coded
- Height adjustable
- Offer complete control over loading volume and well depth
Specifications
HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit | |
28 | |
30 μL |
Comb | |
1.5 mm | |
5 mm |