Learn More
Hoefer™ Combs for HE 33 Mini Submarine Unit
Accessory parts for HE 33 Mini Submarine
530.00 SEK - 835.00 SEK
Product Code | Brand | Type | No. of Wells | Thickness (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | No. of Wells | Thickness (Metric) | Price | Quantity & Availability | |||
10036334
|
Hoefer™
HE31A1215 |
Backless Comb | 12 | 0.15 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11509533
|
Hoefer™
HE31A-8-1.5 |
Backless Comb | 8 | 0.15 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11519533
|
Hoefer™
HE31A-12-1.0 |
Backless Comb | 12 | 0.1 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11559533
|
Hoefer™
HE31A-P-1.0 |
Backless Preparative Comb | Preparative | 0.1 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11539533
|
Hoefer™
HE31A-16-1.0 |
Backless Comb | 16 | 0.1 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11569533
|
Hoefer™
HE31A-P-1.5 |
Backless Preparative Comb | Preparative | 0.15 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12084637
|
Hoefer™
HE31A-12-1.5 |
Backless Comb | 12 | 0.15 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12044817
|
Hoefer™
HE31-BK |
Comb Back and Two Screws | 1 Prep, 2 Reference |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12074827
|
Hoefer™
HE31A-16-1.5 |
Backless Comb | 16 | 0.15 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11599523
|
Hoefer™
HE31A-8-1.0 |
Backless Comb | 8 | 0.1 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Variety of eight sizes ranging from 1/2 to 16 wells
- 1 to 1.5 mm Thickness
- 44/6 to 2.6 mm Well Width
- 235/35 to 20.5 μL Sample Volume
Specifications
HE33 Mini Submarine Unit |