missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU20 Mini-gel Horizontal Electrophoresis Unit
Offers complete control over loading volume and well depth.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- Color coded
- Height adjustable
- For use with HU20 Mini-gel Horizontal Electrophoresis Unit
Specifications
HU20 Mini-gel Horizontal Electrophoresis Unit | |
16 | |
2 mm | |
8.5 mm |
Comb | |
1 Each | |
75 μL |