missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Chest Freezers, 100 mm Box Height
Comfort Rack suitable for use with all chest freezer models.
Brand: Tenak™ TE24560
Additional Details : Weight : 1.04000kg
Description
- Racks with spring locks mounded on each shelf to secure the cryoboxes
- Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
- Measure the inside height of your freezer when choosing the right rack
- Constructed of stainless steel
Specifications
Spring Locks | |
Stainless Steel | |
140 x 140 x 320 mm |
Boxes (3) | |
Chest Frseezers | |
100 mm (Box) |