missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Chest Freezers, 75 mm Box Height

Comfort Rack suitable for use with all chest freezer models.

Brand:  Tenak™ TE24542

Additional Details : Weight : 1.48000kg

 View more versions of this product

Product Code. 15240844

  • 1090.00 SEK / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Racks with spring locks mounded on each shelf to secure the cryoboxes
  • Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
  • Measure the inside height of your freezer when choosing the right rack
  • Constructed of stainless steel
Specifications

Specifications

Spring Locks
Stainless Steel
140 x 140 x 629 mm
Boxes (8)
Chest Frseezers
75 mm (Box)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers