missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Chest Freezers, 75 mm Box Height
Comfort Rack suitable for use with all chest freezer models.
875.00 SEK - 1170.00 SEK
Specifications
Includes | Spring Locks |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 75 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15200844
|
Tenak™
TE24538 |
Boxes (6) |
1035.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15220834
|
Tenak™
TE24530 |
Boxes (2) |
875.00 SEK
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15220844
|
Tenak™
TE24540 |
Boxes (7) |
1065.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15240834
|
Tenak™
TE24532 |
Boxes (3) |
925.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15240844
|
Tenak™
TE24542 |
Boxes (8) |
1090.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15260834
|
Tenak™
TE24534 |
Boxes (4) |
950.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15260844
|
Tenak™
TE24544 |
Boxes (9) |
1170.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15280834
|
Tenak™
TE24536 |
Boxes (5) |
975.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
- Racks with spring locks mounded on each shelf to secure the cryoboxes
- Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
- Measure the inside height of your freezer when choosing the right rack
- Constructed of stainless steel
Specifications
Spring Locks | |
Stainless Steel | |
Chest Frseezers | |
75 mm (Box) |