missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Chest Freezers, 75 mm Box Height, with Boxes

Comfort Rack suitable for use with all chest freezer models.

Brand:  Tenak™ TE24530B

Additional Details : Weight : 1.02000kg

 View more versions of this product

Product Code. 15230834

  • 990.00 SEK / Each
Estimated Shipment: 10-07-2024
to see stock.
Add to basket

Description

Description

  • Racks with spring locks mounded on each shelf to secure the cryoboxes
  • Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
  • Measure the inside height of your freezer when choosing the right rack
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

Spring Locks
Stainless Steel
140 x 140 x 166 mm
Boxes (2)
Chest Frseezers
75 mm (Box)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers